Gene Transcripts

Gene Symbol
ALG2
Entrez Gene ID
85365
Gene Name
ALG2, alpha-1,3/1,6-mannosyltransferase
Gene Aliases
CDGIi, CMS14, CMSTA3, NET38, hALPG2
Chromosome Location
Chr. 9: 99216425 - 99221964 on Build GRCh38
UniGene ID
Hs.40919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033087.3 NP_149078.1 2 589
NR_024532.1 3 748
GenBank mRNA AB161356.1 - 2 546
AK027417.1 - 2 541
AK027814.1 - 1 1341
AK074704.1 - 2 541
AK074988.1 - 3 726
AK075172.1 - 2 565
AY358697.1 - 2 333
BC017876.1 - 2 539 4698763

View Full Product Details