Gene Transcripts

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
Chromosome Location
Chr. 5: 119071002 - 119250170 on Build GRCh38
UniGene ID
Hs.181042
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290321.2 NP_001277250.1 9 1462
NM_001290322.2 NP_001277251.1 9 2770
NM_005509.5 NP_005500.4 9 1462
XM_005271909.4 XP_005271966.1 10 1160
XM_005271910.4 XP_005271967.1 9 1505
XM_005271912.2 XP_005271969.1 10 1144
XM_011543212.2 XP_011541514.1 9 1505
XM_011543213.2 XP_011541515.1 10 1090
XM_011543214.2 XP_011541516.1 6 671
XM_011543215.2 XP_011541517.1 5 507
XM_017009141.1 XP_016864630.1 10 1160
XM_017009142.1 XP_016864631.1 10 1090
XM_017009143.1 XP_016864632.1 11 1339
XM_017009144.1 XP_016864633.1 9 1042
XM_017009145.1 XP_016864634.1 9 912
XM_017009146.1 XP_016864635.1 9 2354
XM_017009147.1 XP_016864636.1 9 1505
XM_017009148.1 XP_016864637.1 8 1308
GenBank mRNA AB593142.1 - 9 2773
AJ005821.1 - 9 1059
BC028004.1 - 10 1047
BC144537.1 - 9 1005 9053064
BC150616.1 - 9 1005 9056986

View Full Product Details