Gene Transcripts

Gene Symbol
SEC16B
Entrez Gene ID
89866
Gene Name
SEC16 homolog B, endoplasmic reticulum export factor
Gene Aliases
LZTR2, PGPR-p117, RGPR, SEC16S
Chromosome Location
Chr. 1: 177929107 - 177969915 on Build GRCh38
UniGene ID
Hs.149540
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033127.2 NP_149118.2 2 365
GenBank mRNA AB063357.1 - 2 365
AK314585.1 - 2 431
BC009106.1 - 2 357 3448742
EF125213.1 - 1 275

View Full Product Details