Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052848.1 | NP_443080.1 | 2 | 376 | |
XM_005259377.4 | XP_005259434.1 | 2 | 739 | ||
XM_017027442.1 | XP_016882931.1 | 2 | 710 | ||
GenBank mRNA | AL833726.1 | - | 1 | 211 | |
BC011577.1 | - | 2 | 376 | 4651484 |