Gene Transcripts
- Gene Symbol
- IFT20
- Entrez Gene ID
- 90410
- Gene Name
- intraflagellar transport 20
- Gene Aliases
- Chromosome Location
- Chr. 17: 28328325 - 28335489 on Build GRCh38
- UniGene ID
- Hs.705431
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267774.1 |
NP_001254703.1
|
5, 6 |
715 |
|
NM_001267775.1 |
NP_001254704.1
|
5, 6 |
514 |
|
NM_001267776.1 |
NP_001254705.1
|
4, 5 |
478 |
|
NM_001267778.1 |
NP_001254707.1
|
3, 4 |
374 |
|
NM_174887.3 |
NP_777547.1
|
5, 6 |
596 |
|
GenBank mRNA |
AA490722.1 |
- |
3, 4 |
290 |
|
AF070643.1 |
- |
1 |
1086 |
|
AY224601.1 |
- |
5, 6 |
664 |
|
BC002640.2 |
- |
5, 6 |
576 |
3609552 |
BC038094.1 |
- |
4, 5 |
444 |
3907361 |
BE619475.1 |
- |
5, 6 |
450 |
|
View Full Product Details