Gene Transcripts

Gene Symbol
SLC39A13
Entrez Gene ID
91252
Gene Name
solute carrier family 39 member 13
Gene Aliases
LZT-Hs9
Chromosome Location
Chr. 11: 47407132 - 47416500 on Build GRCh38
UniGene ID
Hs.523664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128225.2 NP_001121697.1 5, 6 822
NM_152264.4 NP_689477.2 5, 6 843
NR_134854.1 5, 6 912
XM_006718381.3 XP_006718444.1 5, 6 727
XM_006718384.2 XP_006718447.1 5, 6 727
XM_011520466.1 XP_011518768.1 5, 6 727
XM_011520467.1 XP_011518769.1 5, 6 1056
XM_011520468.2 XP_011518770.1 5, 6 1149
XM_017018540.1 XP_016874029.1 5, 6 701
XM_017018542.1 XP_016874031.1 5, 6 701
GenBank mRNA AK098651.1 - 5, 6 912
BC008853.2 - 5, 6 688 4100291
BC019016.2 - 5, 6 707 4100057
BI918365.1 - 4, 5 404
BX342307.2 - 5, 6 821
BX647491.1 - 7, 8 1007
CF132216.1 - 4, 5 429

View Full Product Details