Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172309.1 | NP_001165780.1 | 11 | 1621 | |
NM_144573.3 | NP_653174.3 | 12 | 1813 | ||
XM_005271322.3 | XP_005271379.1 | 12 | 1704 | ||
XM_005271323.3 | XP_005271380.1 | 11 | 1662 | ||
XM_005271324.4 | XP_005271381.1 | 11 | 1608 | ||
XM_005271325.3 | XP_005271382.1 | 11 | 1482 | ||
XM_005271326.3 | XP_005271383.1 | 10 | 1497 | ||
XM_005271327.3 | XP_005271384.1 | 9 | 1287 | ||
GenBank mRNA | AF114264.1 | - | 8 | 1243 | |
AK057954.1 | - | 8 | 1035 | ||
AK127535.1 | - | 4 | 1090 | ||
AK293201.1 | - | 5 | 726 | ||
AK298565.1 | - | 11 | 1590 | ||
AK300728.1 | - | 11 | 1704 | ||
AK307487.1 | - | 10 | 1497 | ||
BC111395.1 | - | 8 | 1202 | 4271572 | |
BC114444.1 | - | 8 | 1219 | 40083839 | |
BC114445.1 | - | 8 | 1219 | 40083847 | |
DQ464902.1 | - | 12 | 1700 |