Gene Transcripts

Gene Symbol
ORMDL3
Entrez Gene ID
94103
Gene Name
ORMDL sphingolipid biosynthesis regulator 3
Gene Aliases
Chromosome Location
Chr. 17: 39921041 - 39927804 on Build GRCh38
UniGene ID
Hs.514151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320801.1 NP_001307730.1 6 1843
NM_001320802.1 NP_001307731.1 4 681
NM_001320803.1 NP_001307732.1 4 395
NM_139280.3 NP_644809.1 4 686
XM_017025345.1 XP_016880834.1 4 539
XM_017025346.1 XP_016880835.1 4 452
XM_017025347.1 XP_016880836.1 4 632
XM_017025348.1 XP_016880837.1 4 378
GenBank mRNA AF373101.1 - 4 422
AF373102.1 - 4 492
AF395708.1 - 4 477
AK074811.1 - 4 483
AK075212.1 - 4 438
AK093063.1 - 6 1838
AX775843.1 - 4 440
AY357943.1 - 3 295
BC017087.1 - 4 452 3850211
BC071833.1 - 4 431 6340437
DB042662.1 - 4 396
DB127807.1 - 4 514

View Full Product Details