Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003712.1 | NP_001003712.1 | 20 | 2698 | |
NM_001319652.1 | NP_001306581.1 | 23 | 2823 | ||
NM_001319653.1 | NP_001306582.1 | 21 | 2735 | ||
NM_001319655.1 | NP_001306584.1 | 18 | 2629 | ||
NM_020841.4 | NP_065892.1 | 21 | 2744 | ||
XM_005268621.4 | XP_005268678.1 | 24 | 2842 | ||
XM_006719224.2 | XP_006719287.1 | 20 | 2703 | ||
XM_011537855.2 | XP_011536157.1 | 21 | 2361 | ||
XM_011537856.2 | XP_011536158.1 | 22 | 2779 | ||
XM_011537857.2 | XP_011536159.1 | 24 | 2833 | ||
XM_017018768.1 | XP_016874257.1 | 21 | 2376 | ||
XM_017018769.1 | XP_016874258.1 | 22 | 2747 | ||
GenBank mRNA | AB040884.3 | - | 21 | 2415 | |
AF392452.1 | - | 21 | 2745 | ||
AK172813.1 | - | 3 | 244 | ||
AK289997.1 | - | 21 | 2464 | ||
AL832876.1 | - | 8 | 800 | ||
BC012935.1 | - | 7 | 729 | ||
BC093834.1 | - | 20 | 2343 | 7939679 | |
BC101529.1 | - | 20 | 2343 | 8069035 | |
BC111728.1 | - | 21 | 2638 | 40029118 | |
BX647211.1 | - | 20 | 2300 | ||
CR749542.1 | - | 23 | 2548 |