Gene Transcripts
- Gene Symbol
- UHRF2
- Entrez Gene ID
- 115426
- Gene Name
- ubiquitin like with PHD and ring finger domains 2
- Gene Aliases
- NIRF, RNF107, TDRD23, URF2
- Chromosome Location
- Chr. 9: 6413148 - 6508108 on Build GRCh38
- UniGene ID
- Hs.493401
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152896.2 |
NP_690856.1
|
5, 6 |
1309 |
|
NR_046386.1 |
|
5, 6 |
1309 |
|
XM_011517704.2 |
XP_011516006.1
|
5, 6 |
938 |
|
XM_011517705.2 |
XP_011516007.1
|
3, 4 |
437 |
|
XM_017014253.1 |
XP_016869742.1
|
4, 5 |
1093 |
|
XM_017014254.1 |
XP_016869743.1
|
6, 7 |
1041 |
|
GenBank mRNA |
AB071698.1 |
- |
5, 6 |
1007 |
|
AF274049.1 |
- |
5, 6 |
1137 |
|
AK122730.1 |
- |
3, 4 |
411 |
|
AK308433.1 |
- |
5, 6 |
1262 |
|
BC028397.2 |
- |
5, 6 |
1309 |
4797830 |
View Full Product Details