Gene Transcripts

Gene Symbol
CMTM5
Entrez Gene ID
116173
Gene Name
CKLF like MARVEL transmembrane domain containing 5
Gene Aliases
CKLFSF5
Chromosome Location
Chr. 14: 23376439 - 23379772 on Build GRCh38
UniGene ID
Hs.99272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037288.1 NP_001032365.1 4 891
NM_001288744.1 NP_001275673.1 4 831
NM_001288745.1 NP_001275674.1 3 738
NM_001288746.1 NP_001275675.1 6 1185
NM_138460.2 NP_612469.1 5 984
XM_017020953.1 XP_016876442.1 6 970
XM_017020954.1 XP_016876443.1 6 847
GenBank mRNA AF479262.2 - 5 985
AF527413.1 - 6 968
AF527414.1 - 5 615
AF527948.2 - 3 509
AF527949.1 - 4 602
AK094840.1 - 4 1716
BC013109.1 - 5 729 4214683

View Full Product Details