Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135556.1 | NP_001129028.1 | 8 | 941 | |
NM_001135557.1 | NP_001129029.1 | 7 | 881 | ||
NM_001278421.1 | NP_001265350.1 | 7 | 932 | ||
NM_001278422.1 | NP_001265351.1 | 7 | 881 | ||
NM_004411.4 | NP_004402.1 | 8 | 992 | ||
XM_011515861.1 | XP_011514163.1 | 8 | 839 | ||
XM_011515862.1 | XP_011514164.1 | 8 | 788 | ||
XM_011515863.2 | XP_011514165.1 | 8 | 946 | ||
XM_017011804.1 | XP_016867293.1 | 8 | 1579 | ||
XM_017011805.1 | XP_016867294.1 | 8 | 766 | ||
XM_017011806.1 | XP_016867295.1 | 8 | 941 | ||
XM_017011807.1 | XP_016867296.1 | 8 | 946 | ||
GenBank mRNA | AF063228.1 | - | 8 | 902 | |
AF123074.1 | - | 7 | 790 | ||
AK091339.1 | - | 9 | 841 | ||
AK297427.1 | - | 7 | 793 | ||
AK309419.1 | - | 8 | 764 | ||
AK313148.1 | - | 8 | 939 | ||
BC022540.1 | - | 7 | 841 | 4793415 | |
BC032945.2 | - | 8 | 893 | 4801387 | |
BC037854.1 | - | 8 | 883 | 4817657 | |
BC053542.1 | - | 8 | 881 | 5727968 |