Gene Transcripts

Gene Symbol
AGAP11
Entrez Gene ID
119385
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
Gene Aliases
Chromosome Location
Chr. 10: 86970741 - 87010203 on Build GRCh38
UniGene ID
Hs.511787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133447.1 NP_597704.1 11 1274
GenBank mRNA AB075855.1 - 6 2678
AK097165.1 - 11 1274
BC140762.1 - 7 520 9021630

View Full Product Details