Gene Transcripts

Gene Symbol
FAT3
Entrez Gene ID
120114
Gene Name
FAT atypical cadherin 3
Gene Aliases
CDHF15, CDHR10, hFat3
Chromosome Location
Chr. 11: 92224636 - 92896473 on Build GRCh38
UniGene ID
Hs.98523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008781.2 NP_001008781.2 1 3294
XM_017017178.1 XP_016872667.1 2 3410
XM_017017179.1 XP_016872668.1 3 3632
XM_017017180.1 XP_016872669.1 2 3409
XM_017017181.1 XP_016872670.1 2 3409
XM_017017182.1 XP_016872671.1 2 3409
XM_017017183.1 XP_016872672.1 2 3409
XM_017017184.1 XP_016872673.1 2 3408
GenBank mRNA AK054903.1 - 1 2026
BC016722.1 - 1 207

View Full Product Details