Gene Transcripts

Gene Symbol
PIH1D2
Entrez Gene ID
120379
Gene Name
PIH1 domain containing 2
Gene Aliases
Chromosome Location
Chr. 11: 112052161 - 112074288 on Build GRCh38
UniGene ID
Hs.420662
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082619.1 NP_001076088.1 5 790
NM_138789.3 NP_620144.1 5 790
XM_011542593.2 XP_011540895.2 5 696
XM_017017201.1 XP_016872690.1 5 907
XM_017017202.1 XP_016872691.1 5 907
XM_017017203.1 XP_016872692.1 5 907
XM_017017204.1 XP_016872693.1 5 907
XM_017017205.1 XP_016872694.1 5 907
GenBank mRNA AK300496.1 - 5 653
BC019238.2 - 4 599 3935322

View Full Product Details