Gene Transcripts

Gene Symbol
ANO4
Entrez Gene ID
121601
Gene Name
anoctamin 4
Gene Aliases
TMEM16D
Chromosome Location
Chr. 12: 100717457 - 101128641 on Build GRCh38
UniGene ID
Hs.58785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286615.1 NP_001273544.1 22 2594
NM_001286616.1 NP_001273545.1 21 2162
NM_178826.3 NP_849148.2 21 2489
XM_011537911.2 XP_011536213.1 24 2589
XM_011537912.2 XP_011536214.1 24 2589
XM_011537913.2 XP_011536215.1 23 2544
XM_011537914.2 XP_011536216.1 21 2100
XM_011537915.2 XP_011536217.1 22 2412
XM_011537916.2 XP_011536218.1 22 2253
XM_017018808.1 XP_016874297.1 21 2100
XM_017018809.1 XP_016874298.1 23 2680
XM_017018810.1 XP_016874299.1 21 2342
XM_017018811.1 XP_016874300.1 20 2297
XM_017018812.1 XP_016874301.1 20 4315
XM_017018813.1 XP_016874302.1 19 2165
XM_017018814.1 XP_016874303.1 22 4554
XM_017018815.1 XP_016874304.1 22 4554
XM_017018816.1 XP_016874305.1 22 2637
XM_017018817.1 XP_016874306.1 22 4589
GenBank mRNA AK091540.1 - 12 1110
AK091591.1 - 21 2278
AK092596.1 - 9 676
AK293810.1 - 22 2232
AK316105.1 - 22 2233
BC109308.2 - 21 2147 40035759

View Full Product Details