Gene Transcripts

Gene Symbol
SLAIN1
Entrez Gene ID
122060
Gene Name
SLAIN motif family member 1
Gene Aliases
C13orf32
Chromosome Location
Chr. 13: 77697854 - 77764242 on Build GRCh38
UniGene ID
Hs.349955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040153.3 NP_001035243.2 4 558
NM_001242868.1 NP_001229797.1 4 1067
NM_001242871.1 NP_001229800.1 3 606
NM_144595.3 NP_653196.1 3 522
XM_011534929.2 XP_011533231.1 5 947
XM_011534930.1 XP_011533232.1 4 460
GenBank mRNA AK054608.1 - 3 302
AK289714.1 - 3 312
AK294201.1 - 4 558
AK295423.1 - 3 382
AL834332.1 - 3 315
BC045177.1 - 3 331
BC111497.1 - 3 313 6148614
BG391962.1 - 446
DA343547.1 - 3 522

View Full Product Details