Gene Transcripts
- Gene Symbol
- C14orf79
- Entrez Gene ID
- 122616
- Gene Name
- chromosome 14 open reading frame 79
- Gene Aliases
- Chromosome Location
- Chr. 14: 104985801 - 105000310 on Build GRCh38
- UniGene ID
- Hs.27183
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_174891.3 |
NP_777551.2
|
3, 4 |
849 |
|
XM_005267317.3 |
XP_005267374.1
|
3, 4 |
1327 |
|
XM_005267318.3 |
XP_005267375.1
|
3, 4 |
1327 |
|
GenBank mRNA |
AF038188.1 |
- |
3, 4 |
413 |
|
AK057249.1 |
- |
4, 5 |
925 |
|
BC003539.1 |
- |
2, 3 |
161 |
|
BC011537.1 |
- |
2, 3 |
679 |
|
BC025175.1 |
- |
3, 4 |
360 |
|
BC137495.1 |
- |
3, 4 |
792 |
9021495 |
BC144671.1 |
- |
3, 4 |
778 |
|
View Full Product Details