Gene Transcripts

Gene Symbol
FRMD6
Entrez Gene ID
122786
Gene Name
FERM domain containing 6
Gene Aliases
C14orf31, EX1, Willin, c14_5320
Chromosome Location
Chr. 14: 51489100 - 51730727 on Build GRCh38
UniGene ID
Hs.434914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042481.2 NP_001035946.1 15 2285
NM_001267046.1 NP_001253975.1 14 2085
NM_001267047.1 NP_001253976.1 5 842
NM_152330.3 NP_689543.1 14 2061
XM_006720029.1 XP_006720092.1 14 2182
XM_006720030.1 XP_006720093.1 14 2158
XM_011536423.1 XP_011534725.1 15 2329
XM_011536424.1 XP_011534726.1 16 2454
GenBank mRNA AK055545.1 - 14 1996
AK126521.1 - 12 1706
AK129995.1 - 1 157
AK315764.1 - 14 1982
AL833158.1 - 15 2271
AL833380.1 - 9 1300
AL833661.1 - 9 1281
BC020521.2 - 14 2009 3914276
BC029870.1 - 5 842
BX161430.1 - 14 2061
BX647239.1 - 5 2630
BX647863.1 - 14 1973
BX648295.1 - 5 2631

View Full Product Details