Gene Transcripts

Gene Symbol
SPATA33
Entrez Gene ID
124045
Gene Name
spermatogenesis associated 33
Gene Aliases
C16orf55
Chromosome Location
Chr. 16: 89657744 - 89670458 on Build GRCh38
UniGene ID
Hs.655171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271907.1 NP_001258836.1 2 175
NM_001271908.1 NP_001258837.1 2 93
NM_001271909.1 NP_001258838.1 1 231
NM_001271910.1 NP_001258839.1 2 175
NM_153025.2 NP_694570.1 2 122
NR_073516.1 1 231
XM_006721133.2 XP_006721196.1 2 175
GenBank mRNA AK056168.1 - 2 64
AK303024.1 - 2 141
BC148299.2 - 2 64 40112476
BP345094.1 - 1 231
DA300380.1 - 2 85
DB025192.1 - 2 122
DB452323.1 - 2 175
DC357681.1 - 2 133
DC374690.1 - 2 143

View Full Product Details