Gene Transcripts
- Gene Symbol
- SPATA33
- Entrez Gene ID
- 124045
- Gene Name
- spermatogenesis associated 33
- Gene Aliases
- C16orf55
- Chromosome Location
- Chr. 16: 89657744 - 89670458 on Build GRCh38
- UniGene ID
- Hs.655171
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271907.1 |
NP_001258836.1
|
2, 3 |
313 |
|
NM_001271908.1 |
NP_001258837.1
|
2, 3 |
231 |
|
NM_001271909.1 |
NP_001258838.1
|
1, 2 |
369 |
|
NM_153025.2 |
NP_694570.1
|
2, 3 |
260 |
|
GenBank mRNA |
AK056168.1 |
- |
2, 3 |
202 |
|
AK303024.1 |
- |
2, 3 |
279 |
|
BC148299.2 |
- |
2, 3 |
202 |
40112476 |
BP345094.1 |
- |
1, 2 |
369 |
|
DA300380.1 |
- |
2, 3 |
223 |
|
DB025192.1 |
- |
2, 3 |
260 |
|
DB452323.1 |
- |
2, 3 |
313 |
|
DC374690.1 |
- |
2, 3 |
281 |
|
View Full Product Details