Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136052.2 | NP_001129524.1 | 4 | 563 | |
NM_001199752.2 | NP_001186681.1 | 4 | 627 | ||
NM_001199753.1 | NP_001186682.1 | 4 | 563 | ||
NM_152359.2 | NP_689572.1 | 3 | 494 | ||
NR_108072.1 | 5 | 725 | |||
XM_005258505.2 | XP_005258562.1 | 4 | 600 | ||
XM_005258506.4 | XP_005258563.1 | 3 | 516 | ||
XM_006723009.3 | XP_006723072.1 | 3 | 405 | ||
XM_011526438.2 | XP_011524740.1 | 4 | 656 | ||
XM_011526439.2 | XP_011524741.1 | 4 | 720 | ||
XM_011526440.2 | XP_011524742.1 | 4 | 572 | ||
XM_017026265.1 | XP_016881754.1 | 4 | 619 | ||
XM_017026266.1 | XP_016881755.1 | 4 | 573 | ||
XM_017026267.1 | XP_016881756.1 | 4 | 562 | ||
XM_017026268.1 | XP_016881757.1 | 3 | 405 | ||
XM_017026269.1 | XP_016881758.1 | 3 | 515 | ||
XM_017026270.1 | XP_016881759.1 | 3 | 283 | ||
XM_017026271.1 | XP_016881760.1 | 3 | 405 | ||
XM_017026272.1 | XP_016881761.1 | 4 | 599 | ||
XM_017026273.1 | XP_016881762.1 | 4 | 584 | ||
GenBank mRNA | AF357970.1 | - | 3 | 452 | |
AK096516.1 | - | 5 | 721 | ||
AK289713.1 | - | 4 | 619 | ||
AK290092.1 | - | 4 | 535 | ||
AK299866.1 | - | 3 | 394 | ||
AK309087.1 | - | 4 | 555 | ||
BC029104.1 | - | 4 | 527 | 3872727 |