Gene Transcripts

Gene Symbol
NLRP8
Entrez Gene ID
126205
Gene Name
NLR family pyrin domain containing 8
Gene Aliases
CLR19.2, NALP8, NOD16, PAN4
Chromosome Location
Chr. 19: 55947832 - 55988629 on Build GRCh38
UniGene ID
Hs.446925
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317000.1 NP_001303929.1 3 1745
NM_176811.2 NP_789781.2 3 1745
GenBank mRNA AY154463.1 - 3 1674

View Full Product Details