Gene Transcripts

Gene Symbol
CCSAP
Entrez Gene ID
126731
Gene Name
centriole, cilia and spindle associated protein
Gene Aliases
C1orf96, CSAP
Chromosome Location
Chr. 1: 229321005 - 229342941 on Build GRCh38
UniGene ID
Hs.585011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145257.4 NP_660300.3 3 549
GenBank mRNA AK123465.1 - 2 338
AK291437.1 - 2 510
BC015419.1 - 2 523 4396825
BC060777.1 - 3 1454
BC071609.1 - 3 595 5264741

View Full Product Details