Gene Transcripts

Gene Symbol
LRRC38
Entrez Gene ID
126755
Gene Name
leucine rich repeat containing 38
Gene Aliases
Chromosome Location
Chr. 1: 13474977 - 13513747 on Build GRCh38
UniGene ID
Hs.657356
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010847.1 NP_001010847.1 2 1522
GenBank mRNA BC016048.1 - 2 1437

View Full Product Details