Gene Transcripts
- Gene Symbol
- OR2M5
- Entrez Gene ID
- 127059
- Gene Name
- olfactory receptor family 2 subfamily M member 5
- Gene Aliases
- OR2M5P
- Chromosome Location
- Chr. 1: 248145148 - 248146086 on Build GRCh38
- UniGene ID
- Hs.553580
- Species
- Homo sapiens
- Gene Symbol
- OR2M1P
- Entrez Gene ID
- 388762
- Gene Name
- olfactory receptor family 2 subfamily M member 1 pseudogene
- Gene Aliases
- JCG10, OR2M1, OST037
- Chromosome Location
- Chr. 1: 248122136 - 248122780 on Build GRCh38
- UniGene ID
- Hs.528509
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002141.1 |
|
1 |
564 |
|
View Full Product Details