Gene Transcripts

Gene Symbol
HMGB4
Entrez Gene ID
127540
Gene Name
high mobility group box 4
Gene Aliases
dJ1007G16.5
Chromosome Location
Chr. 1: 33860475 - 33864791 on Build GRCh38
UniGene ID
Hs.568628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145205.5 NP_660206.2 2 2217
NR_033264.1 2 502
GenBank mRNA AK097707.1 - 2 2218
AK130859.1 - 1 636
AK310465.1 - 2 502
AK311985.1 - 1 603
BC021180.2 - 1 587 5266418
BC070148.1 - 2 502 6617009
BI827031.1 - 1 589
DQ207367.1 - 1 587

View Full Product Details