Gene Transcripts

Gene Symbol
OR9A4
Entrez Gene ID
130075
Gene Name
olfactory receptor family 9 subfamily A member 4
Gene Aliases
Chromosome Location
Chr. 7: 141918876 - 141919820 on Build GRCh38
UniGene ID
Hs.690171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001656.1 NP_001001656.1 1 504
GenBank mRNA BC136819.1 - 1 563
BC136821.1 - 1 563 9020812

View Full Product Details