Gene Transcripts
- Gene Symbol
- UBR3
- Entrez Gene ID
- 130507
- Gene Name
- ubiquitin protein ligase E3 component n-recognin 3 (putative)
- Gene Aliases
- ZNF650
- Chromosome Location
- Chr. 2: 169827463 - 170084131 on Build GRCh38
- UniGene ID
- Hs.379548
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172070.3 |
NP_742067.3
|
34, 35 |
4894 |
|
XM_005246299.4 |
XP_005246356.1
|
35, 36 |
4984 |
|
XM_005246300.4 |
XP_005246357.1
|
35, 36 |
4927 |
|
XM_006712268.3 |
XP_006712331.1
|
35, 36 |
4981 |
|
XM_006712269.3 |
XP_006712332.1
|
34, 35 |
4906 |
|
XM_006712270.3 |
XP_006712333.2
|
34, 35 |
4840 |
|
XM_006712271.3 |
XP_006712334.1
|
34, 35 |
4947 |
|
GenBank mRNA |
AB095944.1 |
- |
5, 6 |
1004 |
|
AK126998.1 |
- |
12, 13 |
1623 |
|
AK127553.1 |
- |
8, 9 |
900 |
|
AK128490.1 |
- |
18, 19 |
2374 |
|
AK303062.1 |
- |
9, 10 |
994 |
|
AL834144.1 |
- |
6, 7 |
601 |
|
BX649036.1 |
- |
10, 11 |
1130 |
|
CR749469.1 |
- |
12, 13 |
1396 |
|
View Full Product Details