Gene Transcripts

Gene Symbol
UBR3
Entrez Gene ID
130507
Gene Name
ubiquitin protein ligase E3 component n-recognin 3 (putative)
Gene Aliases
ZNF650
Chromosome Location
Chr. 2: 169827463 - 170084131 on Build GRCh38
UniGene ID
Hs.379548
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172070.3 NP_742067.3 31 4490
XM_005246299.4 XP_005246356.1 32 4580
XM_005246300.4 XP_005246357.1 32 4523
XM_006712268.3 XP_006712331.1 32 4577
XM_006712269.3 XP_006712332.1 31 4502
XM_006712270.3 XP_006712333.2 31 4436
XM_006712271.3 XP_006712334.1 31 4543
XM_017003356.1 XP_016858845.1 32 4535
GenBank mRNA AB095944.1 - 2 600
AK126998.1 - 9 1219
AK127553.1 - 5 496
AK128490.1 - 15 1970
AK303062.1 - 6 590
AL834144.1 - 3 197
BX649036.1 - 7 726
CR749469.1 - 9 992

View Full Product Details