Gene Transcripts

Gene Symbol
ZPLD1
Entrez Gene ID
131368
Gene Name
zona pellucida like domain containing 1
Gene Aliases
Chromosome Location
Chr. 3: 102385144 - 102479841 on Build GRCh38
UniGene ID
Hs.352213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175056.1 NP_778226.1 3 299
XM_017005703.1 XP_016861192.1 4 481
XM_017005704.1 XP_016861193.1 3 337
GenBank mRNA AY090780.1 - 3 299
BC031261.1 - 10 1033 5273136
BC143817.1 - 4 395

View Full Product Details