Gene Transcripts

Gene Symbol
LYZL4
Entrez Gene ID
131375
Gene Name
lysozyme like 4
Gene Aliases
LYC4, LYZA
Chromosome Location
Chr. 3: 42360767 - 42427911 on Build GRCh38
UniGene ID
Hs.234895
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304386.1 NP_001291315.1 3, 4 557
NM_144634.3 NP_653235.1 3, 4 576
XM_011533355.2 XP_011531657.1 3, 4 619
XM_017005705.1 XP_016861194.1 5, 6 931
XM_017005706.1 XP_016861195.1 4, 5 689
GenBank mRNA AF326749.1 - 3, 4 505
BC016747.2 - 3, 4 547 4830396

View Full Product Details