Gene Transcripts

Gene Symbol
RTP1
Entrez Gene ID
132112
Gene Name
receptor transporter protein 1
Gene Aliases
Z3CXXC1
Chromosome Location
Chr. 3: 187197486 - 187201465 on Build GRCh38
UniGene ID
Hs.518480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153708.2 NP_714919.2 1 93
GenBank mRNA BC034744.1 - 1 59
BC065202.1 - 1 93 5179096

View Full Product Details