Gene Transcripts

Gene Symbol
C4orf36
Entrez Gene ID
132989
Gene Name
chromosome 4 open reading frame 36
Gene Aliases
Chromosome Location
Chr. 4: 86876205 - 86897256 on Build GRCh38
UniGene ID
Hs.339646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144645.3 NP_653246.1 4, 5 664
GenBank mRNA BC016746.1 - 4, 5 640
BX090513.1 - 3, 4 405

View Full Product Details