Gene Transcripts

Gene Symbol
TRIM40
Entrez Gene ID
135644
Gene Name
tripartite motif containing 40
Gene Aliases
RNF35
Chromosome Location
Chr. 6: 30135998 - 30148773 on Build GRCh38
UniGene ID
Hs.509439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286633.1 NP_001273562.1 6 1091
NM_138700.4 NP_619645.1 5 922
XM_011514305.1 XP_011512607.1 6 1522
XM_011514306.1 XP_011512608.1 7 1388
XM_011514309.1 XP_011512611.1 5 1499
GenBank mRNA AF489517.1 - 5 645
BC060785.1 - 6 1091 30340682

View Full Product Details