Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004422.2 | NP_004413.1 | 7 | 1090 | |
XM_005256502.1 | XP_005256559.1 | 7 | 1082 | ||
XM_017024298.1 | XP_016879787.1 | 4 | 436 | ||
GenBank mRNA | AF006012.1 | - | 7 | 814 | |
AK026019.1 | - | 6 | 616 | ||
AK297099.1 | - | 7 | 846 | ||
AK297285.1 | - | 7 | 823 | ||
AK304229.1 | - | 7 | 857 | ||
AK312829.1 | - | 7 | 1091 | ||
BC014844.1 | - | 7 | 903 | 3852554 | |
BT009822.1 | - | 7 | 808 |