Gene Transcripts

Gene Symbol
SSMEM1
Entrez Gene ID
136263
Gene Name
serine rich single-pass membrane protein 1
Gene Aliases
C7orf45
Chromosome Location
Chr. 7: 130205399 - 130216869 on Build GRCh38
UniGene ID
Hs.351816
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145268.3 NP_660311.1 1 151
GenBank mRNA AK097635.1 - 1 137
BC017587.2 - 1 155 4838989

View Full Product Details