Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001139456.1 | NP_001132928.1 | 6 | 519 | |
NM_174959.3 | NP_777619.1 | 2 | 136 | ||
XM_005250143.3 | XP_005250200.1 | 7 | 923 | ||
XM_011515797.2 | XP_011514099.1 | 3 | 210 | ||
XM_017011746.1 | XP_016867235.1 | 4 | 325 | ||
XM_017011747.1 | XP_016867236.1 | 3 | 150 | ||
XM_017011748.1 | XP_016867237.1 | 3 | 155 | ||
GenBank mRNA | BC036796.1 | - | 2 | 136 | |
BX648789.1 | - | 2 | 3756 |