Gene Transcripts

Gene Symbol
VPS37A
Entrez Gene ID
137492
Gene Name
VPS37A, ESCRT-I subunit
Gene Aliases
HCRP1, PQBP2, SPG53
Chromosome Location
Chr. 8: 17246892 - 17333532 on Build GRCh38
UniGene ID
Hs.343873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145152.1 NP_001138624.1 2 545
NM_152415.2 NP_689628.2 3 620
XM_005273400.2 XP_005273457.1 4 630
XM_005273401.2 XP_005273458.1 3 576
XM_006716286.1 XP_006716349.1 3 585
XM_017013021.1 XP_016868510.1 3 585
XM_017013022.1 XP_016868511.1 3 585
XM_017013023.1 XP_016868512.1 4 630
XM_017013024.1 XP_016868513.1 3 576
XM_017013025.1 XP_016868514.1 4 630
GenBank mRNA AF547097.1 - 2 314
AK057204.1 - 3 541
AY033079.2 - 3 418
BC067754.1 - 3 533 5275060

View Full Product Details