Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009609.3 | NP_001009609.2 | 2 | 347 | |
XM_017029264.1 | XP_016884753.1 | 2 | 467 | ||
GenBank mRNA | BC062750.1 | - | 351 | ||
DQ336128.1 | - | 2 | 263 | ||
DQ336129.1 | - | 2 | 263 |