Gene Transcripts

Gene Symbol
OTUD6A
Entrez Gene ID
139562
Gene Name
OTU deubiquitinase 6A
Gene Aliases
DUBA2, HSHIN6
Chromosome Location
Chr. X: 70062491 - 70064179 on Build GRCh38
UniGene ID
Hs.447381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207320.2 NP_997203.1 1 240
GenBank mRNA AK098697.1 - 1 240
BC137355.1 - 1 236 9021353
BC137356.1 - 1 236 9021354

View Full Product Details