Gene Transcripts
- Gene Symbol
- PNCK
- Entrez Gene ID
- 139728
- Gene Name
- pregnancy up-regulated nonubiquitous CaM kinase
- Gene Aliases
- BSTK3, CaMK1b
- Chromosome Location
- Chr. X: 153669723 - 153687568 on Build GRCh38
- UniGene ID
- Hs.436667
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039582.3 |
NP_001034671.3
|
3, 4 |
630 |
|
NM_001135740.1 |
NP_001129212.1
|
3, 4 |
264 |
|
XM_005274651.2 |
XP_005274708.1
|
3, 4 |
247 |
|
XM_006724809.3 |
XP_006724872.1
|
3, 4 |
456 |
|
XM_011531107.2 |
XP_011529409.2
|
3, 4 |
335 |
|
XM_011531108.2 |
XP_011529410.1
|
3, 4 |
300 |
|
XM_011531110.2 |
XP_011529412.1
|
3, 4 |
352 |
|
XM_011531111.2 |
XP_011529413.1
|
3, 4 |
270 |
|
XM_011531112.2 |
XP_011529414.1
|
3, 4 |
243 |
|
XM_017029277.1 |
XP_016884766.1
|
3, 4 |
555 |
|
XM_017029278.1 |
XP_016884767.1
|
4, 5 |
459 |
|
XM_017029279.1 |
XP_016884768.1
|
3, 4 |
216 |
|
XM_017029280.1 |
XP_016884769.1
|
3, 4 |
247 |
|
GenBank mRNA |
AK296067.1 |
- |
2, 3 |
551 |
|
AK296165.1 |
- |
3, 4 |
485 |
|
AK296203.1 |
- |
3, 4 |
630 |
|
AK303746.1 |
- |
3, 4 |
264 |
|
AK309148.1 |
- |
3, 4 |
579 |
|
AK316402.1 |
- |
2, 3 |
800 |
|
BC064422.1 |
- |
3, 4 |
215 |
5751849 |
View Full Product Details