Gene Transcripts

Gene Symbol
SPIN4
Entrez Gene ID
139886
Gene Name
spindlin family member 4
Gene Aliases
TDRD28
Chromosome Location
Chr. X: 63347228 - 63351339 on Build GRCh38
UniGene ID
Hs.612782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012968.2 NP_001012986.2 1 1045
GenBank mRNA AK126931.1 - 1 1022
AL833314.1 - 1 1038
BC043412.1 - 1 556 6156224
BC112360.1 - 1 796 40032302

View Full Product Details