Gene Transcripts

Gene Symbol
MYO3B
Entrez Gene ID
140469
Gene Name
myosin IIIB
Gene Aliases
Chromosome Location
Chr. 2: 170178145 - 170655325 on Build GRCh38
UniGene ID
Hs.671900
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083615.3 NP_001077084.2 2 266
NM_138995.4 NP_620482.3 2 266
NR_045682.1 2 266
NR_045683.1 2 266
NR_045684.1 2 266
XM_006712299.3 XP_006712362.1 2 154
XM_011510654.2 XP_011508956.1 2 154
XM_011510655.2 XP_011508957.1 2 154
GenBank mRNA AF369908.1 - 2 266
AF391554.1 - 2 266
AF391555.1 - 2 266
AF391556.1 - 2 266
AF391557.1 - 2 266
AF391558.1 - 2 266
AK055778.1 - 2 154
BC040019.1 - 1 121
BC136620.1 - 2 203 9020610
BC144307.1 - 2 203 9052827
BX647895.1 - 1 77

View Full Product Details