Gene Transcripts

Gene Symbol
SPTY2D1
Entrez Gene ID
144108
Gene Name
SPT2 chromatin protein domain containing 1
Gene Aliases
Spt2
Chromosome Location
Chr. 11: 18606401 - 18634823 on Build GRCh38
UniGene ID
Hs.738650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194285.2 NP_919261.2 5 2125
XM_011519919.2 XP_011518221.1 5 1983
GenBank mRNA AF452716.1 - 3 1519
AK096694.1 - 1 29
AL834393.1 - 2 175
BC036844.1 - 3 745
BX538046.1 - 3 504
BX641102.1 - 5 2026
BX647798.1 - 5 2125
BX648114.1 - 3 1704
BX648478.1 - 5 2047
CR749626.1 - 3 1568

View Full Product Details