Gene Transcripts
- Gene Symbol
- KIF18B
- Entrez Gene ID
- 146909
- Gene Name
- kinesin family member 18B
- Gene Aliases
- Chromosome Location
- Chr. 17: 44924711 - 44947746 on Build GRCh38
- UniGene ID
- Hs.135094
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001264573.1 |
NP_001251503.1
|
5, 6 |
792 |
|
NM_001265577.1 |
NP_001252506.1
|
5, 6 |
792 |
|
XM_011524385.2 |
XP_011522687.1
|
5, 6 |
1093 |
|
XM_011524386.2 |
XP_011522688.1
|
5, 6 |
827 |
|
XM_011524387.2 |
XP_011522689.1
|
5, 6 |
1094 |
|
XM_011524388.2 |
XP_011522690.1
|
5, 6 |
1093 |
|
XM_011524389.2 |
XP_011522691.1
|
5, 6 |
1093 |
|
XM_011524390.2 |
XP_011522692.1
|
4, 5 |
989 |
|
XM_011524391.2 |
XP_011522693.1
|
5, 6 |
1096 |
|
GenBank mRNA |
BC136590.1 |
- |
4, 5 |
718 |
|
BC144271.1 |
- |
4, 5 |
718 |
9052791 |
BX405609.2 |
- |
5, 6 |
790 |
|
GU808359.1 |
- |
4, 5 |
721 |
|
View Full Product Details