Gene Transcripts

Gene Symbol
SLC30A7
Entrez Gene ID
148867
Gene Name
solute carrier family 30 member 7
Gene Aliases
ZNT7, ZnT-7, ZnTL2
Chromosome Location
Chr. 1: 100896076 - 100996913 on Build GRCh38
UniGene ID
Hs.533903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144884.1 NP_001138356.1 1, 2 275
NM_133496.4 NP_598003.2 1, 2 273
XM_011540779.2 XP_011539081.1 1, 2 255
XM_017000400.1 XP_016855889.1 1, 2 273
XM_017000401.1 XP_016855890.1 1, 2 273
GenBank mRNA AF233345.1 - 1, 2 103
AF529197.1 - 1, 2 88
AK313638.1 - 1, 2 256
AY094606.1 - 1, 2 88
BC064692.1 - 1, 2 229 30330665
BX537375.1 - 1, 2 275
CR936806.1 - 1, 2 269
DA997188.1 - 1, 2 273

View Full Product Details