Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136107.1 | NP_001129579.1 | 4 | 716 | |
XM_011509232.2 | XP_011507534.1 | 5 | 737 | ||
XM_011509233.2 | XP_011507535.1 | 6 | 703 | ||
XM_011509234.2 | XP_011507536.1 | 5 | 715 | ||
GenBank mRNA | BC157860.1 | - | 4 | 716 | |
BC171858.1 | - | 4 | 716 | 9054512 |