Gene Transcripts

Gene Symbol
BTLA
Entrez Gene ID
151888
Gene Name
B and T lymphocyte associated
Gene Aliases
BTLA1, CD272
Chromosome Location
Chr. 3: 112458790 - 112499756 on Build GRCh38
UniGene ID
Hs.445162
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085357.1 NP_001078826.1 4 716
NM_181780.3 NP_861445.3 5 860
XM_011512447.2 XP_011510749.1 6 972
XM_017005748.1 XP_016861237.1 6 935
GenBank mRNA AJ717664.1 - 5 657
AK292692.1 - 5 839
AY599411.1 - 4 513
AY606805.1 - 5 657
BC107091.1 - 4 513 40015588
BC107092.1 - 5 655
DQ198368.1 - 4 513

View Full Product Details