Gene Transcripts

Gene Symbol
GLIPR2
Entrez Gene ID
152007
Gene Name
GLI pathogenesis related 2
Gene Aliases
C9orf19, GAPR-1, GAPR1
Chromosome Location
Chr. 9: 36136536 - 36163913 on Build GRCh38
UniGene ID
Hs.493819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287010.1 NP_001273939.1 3 458
NM_001287011.1 NP_001273940.1 3 458
NM_001287013.1 NP_001273942.1 3 295
NM_022343.3 NP_071738.1 3 458
NR_104637.1 5 839
NR_104638.1 3 250
NR_104639.1 3 458
NR_104640.1 3 480
GenBank mRNA AJ011129.1 - 2 215
AK074488.1 - 3 280
AK075091.1 - 3 259
AK091288.1 - 4 1072
AK298741.1 - 3 259
AK307243.1 - 3 259
AY039756.1 - 3 249
BC017918.1 - 3 261 4296721
BF515272.1 - 3 458
CR749781.1 - 5 640
DA268281.1 - 3 295
DA271525.1 - 3 235
DA824243.1 - 3 249
DA986727.1 - 3 259

View Full Product Details